Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MMP17, Recombinant Protein, Invitrogen™

Catalog No. PIRP77535
Click to view available options
Quantity:
10 μg

Recombinant Protein

MMP17 Recombinant Protein, RP-77535, from Invitrogen.

The catalytic domain of human MT4-MMP is produced by tryptic activation of a recombinant soluble proform purified from E. coli periplasm. The polypeptide contains beside the native sequence of 185 amino acids a C-terminal linked polyhistidine-tag. RP-77535 is purified recombinant MMP-17 (catalytic domain) protein, expressed in E.coli. RP-77535 may be used as a control with corresponding anti-MMP-17 antibodies.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9ULZ9
For Use With (Application) Control
Formulation TBS with 0.1% Triton X-100, 5mM CaCl2 and no preservative; pH 7.5
Gene ID (Entrez) 4326
Molecular Weight (g/mol) ∽20
Name Human MMP17
pH Range 7.5
Quantity 10 μg
Source E. coli
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias MT4-MMP
Common Name Human MMP17
Gene Symbol MMP17
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitro research only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.