Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse PMP70 (aa 644-659), Synthetic Peptide, Invitrogen™

Catalog No. PEP038
Click to view available options
Quantity:
50 μg

Synthetic Peptide

Mouse PMP 70 (residues 644-659) Synthetic Peptide, PEP-038, from Invitrogen.

PEP-038 is a 17 amino acid synthetic peptide whose sequence corresponds to carboxy terminus amino acid residues 644-659 of rat PMP70 protein. This sequence is completely conserved in mouse PMP68. The sequence of this peptide is (amino to carboxy terminus): (C)-N644-Y-E-F-K-K-I-T-E-D-T-V-E-F-G-S659 (A cysteine residue was added to facilitate coupling KLH).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and rat PMP70 and mouse PMP68, catalog # PA1-650 and PA1.1-650. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 19299
Species Mouse
Purification Method purified
Accession Number P55096
Common Name Mouse PMP70 (aa 644-659)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Abcd3
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence (C) - N644 - Y - E - F - K - K - I - T - E - D - T - V - E - F - G - S659
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.