Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse SNAP25 (aa 195-206), Synthetic Peptide, Invitrogen™

Catalog No. PEP032
Click to view available options
Quantity:
50 μg

Synthetic Peptide

Mouse SNAP-25 (residues 195-206) Synthetic Peptide, PEP-032, from Invitrogen.

PEP-032 is a 13 amino acid (amino acids 195-206) synthetic peptide whose sequence corresponds to the carboxy terminal of mouse SNAP-25. The sequence of this peptide is (amino to carboxy terminus): (C)-A195-N-Q-R-A-T-K-M-L-G-S-G206 (The N-terminal cysteine has been added to facilitate conjugation and is not a part of the native mouse SNAP-25 sequence)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and SNAP-25 , catalog # PA1-740. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 20614
Species Mouse
Purification Method purified
Accession Number P60879
Common Name Mouse SNAP25 (aa 195-206)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Snap25
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence (C) - A195 - N - Q - R - A - T - K - M - L - G - S - G206
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.