Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Neutrophil elastase, Native Protein, Invitrogen™

Catalog No. PIRP77526
Click to view available options
Quantity:
25 μg

Native Protein

Neutrophil Elastase (human) Native Protein, RP-77526, from Invitrogen.

Leukocyte elastase is one of several hydrolytic enzymes contained in the azurophil granules of human neutrophils exhibiting a relative molecular weight of 29.5 kDa. The enzyme differs from pancreatic elastase in specificity on synthetic substates and in inhibitor sensitivity. Leukozyte elastase shows only moderate sequence homology with pancreatic elastase. It has a very narrow specificity, cleaving Val-X bonds preferentially, and Ala-X links to a lesser extent. It has been shown to degrade elastin, cartilage proteoglycans, several collagens and fibronectin. Physiologically, it is involved in the degradation of foreign materials ingested during phagocytosis. Damage to connective tissue caused by leakage of enzymes is normally limited by proteinase inhibitors. It has been suggested that insufficiant levels of these inhibitors leads to pulmonary emphysema, and elastase in particular has been implicated in abnormal lung connective tissue turnover. RP-77526 is native Neutrophil Elastase protein purified from whole human blood samples. RP-77526 may be used as a control with corresponding anti-Neutrophil Elastase antibodies. RP-77526 is prepared from whole human blood shown to be non-reactive for HbsAG, anti HCV, anti-HBc, and negative for anti-HIV1 2 by FDA approved tests.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08246
Concentration 0.2mg/mL
For Use With (Application) Control
Formulation 0.01M acetic acid with no preservative; pH 5.5
Gene ID (Entrez) 1991
Name Human Neutrophil elastase
pH Range 5.5
Quantity 25 μg
Source Human
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Common Name Human Neutrophil elastase
Gene Symbol ELANE
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species Human
Recombinant Native
Form Liquid
Protein Subtype Native
Purity or Quality Grade purified
Show More Show Less

in vitro research only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.