Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SGMS1, Synthetic Peptide, Invitrogen™
Click to view available options
Quantity:
50 μg
Description
Sphingomyelin Synthase 1 Synthetic Peptide, PEP-12937, from Invitrogen.
Sphingomyelin Synthase is a bidirectional lipid cholinephosphotransferases capable of converting phosphatidylcholine (PC) and ceramide to sphingomyelin (SM) and diacylglycerol (DAG) in addition to performing the inverse reaction. The direction is dependent on the relative concentrations of DAG and ceramide as phosphocholine acceptors. It directly and specifically recognizes the choline head group on the substrate. It also requires two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. It may not function strictly as a SM synthase and is inhibited by bacterial PC-phospholipase C inhibitor D609. It is localized to Golgi apparatus; Golgi membrane; multi-pass membrane protein. It can be possibly present on the brain, heart, kidney, liver, muscle and stomach.
Specifications
Specifications
| Gene ID (Entrez) | 259230 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q86VZ5 |
| Common Name | Human SGMS1 |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with no preservative |
| For Use With (Application) | Control, Western Blot |
| Show More |
in vitro research only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction