Learn More
Thermo Scientific™ QCC (Quick Column Change) for Agilent Instruments
Description
QCC (Quick Column Change)-Leak Free Column Installation
- Tool-free installation of analytical GC column
- Graphite ferrules
Complete QCC Kits Includes:
- Injector Adaptor
- Detector Adaptor
- Tight Nut
- Ferrules (1 each 0.25, 0.32,0.53)
- Ceramic Column Cutter
Specifications
Specifications
| Description | Tight Nut |
| For Use With | Agilent and Thermo Scientific Instruments |