Learn More
Thermo Scientific™ RadEye™ CD System
Contraband and radioactivity detection system ideal for border inspection
| Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
|
19-155-214
|
Thermo Scientific™
425069032 |
Ba-133 source activity: 370 kBq; 10 μCi | N/A | N/A | N/A | ||||
|
19-155-213
|
Thermo Scientific™
425069030 |
Ba-133 source activity: 185 kBq; 5 μCi | N/A | N/A | N/A | ||||
Description
The robust and simple-to-use RadEye CD System for contraband and radioactivity detection is ideal for border inspection. It uses a collimated thin NaI(Tl) detector, a collimated low activity Ba-133 source and a dual channel analyzer.
Key Features and BenefitsDetection of contraband in vehicle cavities