Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Anaerobic Reducible CNA Blood Agar/ Reducible LKV Blood Agar

Catalog No. r02135
Click to view available options
:
Anaerobic Reducible CNA Blood Agar//Reducible LKV Blood Agar
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for the isolation and/or differentiation of microorganisms or specimens

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USDA, APHIS
Manufacturing Origin United States
Media Type Anaerobic reducible CNA Blood Agar/Anaerobic reducible LKV Blood Agar
Formulation Kanamycin <0.1%, Nalidixic Acid <0.1% and Vancomycin <0.1%
For Use With (Application) Isolation and differentiation of microorganisms from samples or specimens
Packaging Type Biplate
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.