Learn More
Thermo Scientific™ Remel™ ImmunoDOT™ Autoimmunity Screening Panel 4 ENA
Catalog No. r245425
Click to view available options
Quantity:
25 Tests
This item is not returnable. View return policy
Catalog No. R245425
Supplier: Thermo Scientific™ R245425
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
EIA membrane test for screening and simultaneous detection of autoantibodies against nuclear antigens in serum and whole blood (SS-A, SS-B, RNP, Sm)
GenBio
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Human Thyroglobulin Autoantibodies, Human Thyroid Peroxidase Autoantibodies
Type Auto Panel 4
Quantity 25 Tests
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.