Learn More
Thermo Scientific™ Remel™ Nutrient Agar
Catalog No. r09627
Click to view available options
:
22mL; Pour tube; 100/Pk
This item is not returnable. View return policy
This item is not returnable. View return policy
General purpose medium for cultivation of less fastidious microorganisms
Nutrient Agar is used for the cultivation of bacteria and for the enumeration of organisms in water, sewage, feces and other materials.
Compliance
Industrial Reference: AOAC, BAM, COMPF, SMD.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.