Learn More
Thermo Scientific™ Remel™ Tryptic Soy Broth (TSB)
Catalog No. R112840
Click to view available options
Quantity:
90 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
(TSB) General purpose enrichment broth for cultivation and isolation of microorganisms
  • Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia) and for use as general purpose enrichment broth
Compliance
Industrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 90 mL
REF numbers beginning with R11 are not intended for IVD use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.