Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tryptic Soy Broth with 6.5% NaCl

Catalog No. r065032
Click to view available options
Quantity:
20 x 5 mL
100 x 5 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Differentiate enterococci from group D streptococci using Tryptic Soy Broth with 6.5% NaCl.

Differentiate enterococci from group D streptococci using Thermo Scientific™ Remel™ Tryptic Soy Broth with 6.5% NaCl. Tryptic Soy Broth (TSB) was originally developed without the addition of serum or blood1 for susceptibility testing of pneumococci to sulfonamides. In 1973, Facklam recommended use of TSB w/6.5% NaCl in combination with the bile esculin test to differentiate nonenterococcal group D streptococci from Enterococcus spp2. Enterococci are salt tolerant and will grow in TSB w/6.5% NaCl while nonenterococcal group D streptococci will not grow in salt broth3.

Used for differentiation between salt-tolerant and salt-intolerant species.

  • Ready-to-use – Convenience of prepared medium
  • Differential medium Enterococcus spp. will grow in the
TRUSTED_SUSTAINABILITY

Specifications

Description Tryptic Soy Broth w/6.5% NaCl
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.