Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Revco Freezer Low-Voltage Safeguards

Catalog No. 1399046A
Click to view available options
:
115V 60Hz

For Thermo Scientific Revco Elite™ and Value™ Freezers

  • Helps protect electrical equipment from damage by automatically boosting low voltage
  • Eliminates start-up problems
  • Provided with terminal board for direct wiring

Order Info

Freestanding; must be field installed by customer .Specify refrigerator or freezer model number and voltage when ordering.

TRUSTED_SUSTAINABILITY

Specifications

Type Low Voltage Safeguard
For Use With (Equipment) Thermo Scientific™ Revco™ Elite™ and Value™ freezer
Voltage 115 V
Electrical Requirements 115 V 60 Hz
Frequency 60 Hz
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.