Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Shandon™ Electronic Floor Scale
Shop All Epredia Products
Click to view available options
Type:
Digital Indicator
Floor Scale
Quick Pit Frame
Description
- A complete line of of epoxy-painted mild steel, electronic floor scales are available
- Variety of sizes are available from 3 × 3 ft. (91.4 × 91.4cm) to 5 × 7 ft. (152.4 × 213.4cm)
- Ramped models are also available
- Variety of capacities are available from 1000 to 10,000 lb. (453.6 to 4536kg)
- No mechanical adjustment is required
- Hermetically sealed load-sensing elements allow the user to clean the unit without damaging the weighing mechanisms
- Standard scale is installed into a pit, but ramped and freestanding units are available
- Pimarily constructed of stainless steel, including the load cells, weighbridge, scale frame, optional ramps
Order Info
Scale 20560 does not include pit frame, ramp, or digital indicator (sold separately)
Certifications
A Certificate of Conformance, Number 91-097 was issued under the National Type Evaluation Program (NTEP).
Compliance
Scales meet /exceed the Class III, 5000 division accuracy with the National Institute of Standards and Technology (NIST™) Handbook 44.
Specifications
Specifications
| Certifications/Compliance | NTEP, meet /exceed the Class 3, 5000 division accuracy with the NIST handbook 44 |
| Material | Stainless Steel, Epoxy |
| Type | Quick Pit Frame |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction