Learn More
Epredia™ Shandon™ HEPA Bone Dust Collector Nozzle Adapters
Fit HEPA Bone Dust Collector to an autopsy saw
Supplier: Epredia™ 1000004
Requires :
- Lipshaw nozzle adapter: 105461 and 105468 necessary for complete assembly.
- 10000/Stryker 90° nozzle adapters: 1000004 and 10015 necessary for complete assembly.
Description
- Nozzle adapter (105462) for Stryker™ Autopsy Saw replaces standard HEPA Bone Dust Collector nozzle
- Bracket extension (10014) for Stryker Autopsy Saw attaches standard nozzle to saw
Order Info
- Shandon 450 Nozzle Adapter 105461 and 105468 necessary for complete assembly. Shandon 10000/Stryker 90° nozzle adapters 1000004 and 10015 necessary for complete assembly.
Specifications
Nozzle Adapter | |
Stryker Autopsy Saw, Shandon 10000 Autopsy Saw |