Learn More
Epredia™ Shandon™ HEPA Bone Dust Collector Nozzle Adapters
Fit HEPA Bone Dust Collector to an autopsy saw
Requires:
- Lipshaw nozzle adapter: 105461 and 105468 necessary for complete assembly.
- 10000/Stryker 90° nozzle adapters: 1000004 and 10015 necessary for complete assembly.
Catalog Number | Mfr. No. | Product Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
10014
|
Epredia™
10014 |
Bracket Extension | Stryker Autopsy Saw | N/A | N/A | N/A | |||
1000004
|
Epredia™
1000004 |
Nozzle Adapter | Stryker Autopsy Saw, Shandon 10000 Autopsy Saw | N/A | N/A | N/A | |||
105461
|
Epredia™
105461 |
Nozzle Adapter | Shandon 450 Autopsy Saw | N/A | N/A | N/A | |||
Description
- Nozzle adapter (105462) for Stryker™ Autopsy Saw replaces standard HEPA Bone Dust Collector nozzle
- Bracket extension (10014) for Stryker Autopsy Saw attaches standard nozzle to saw
Order Info
- Shandon 450 Nozzle Adapter 105461 and 105468 necessary for complete assembly. Shandon 10000/Stryker 90° nozzle adapters 1000004 and 10015 necessary for complete assembly.