Learn More
Epredia™ Shandon™ Surgical Lighting
Catalog No. 20595 Shop All Epredia Products
Click to view available options
Description:
Ceiling-mount, dual head
Track-mount, dual head
Track-mount, single head
Wall-mount, single head
Epredia™ Shandon™ Surgical Lighting is available in a variety of configurations.
  • Complete line of lights provides surgical-quality lighting for use in applications where high intensities and color correction are needed, but field size requirements are less critical, such as in pathology and animal surgery applications
Specifications
Description Ceiling-mount, dual head
Product Type Surgical Lighting
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.