Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sorvall™ Tube for Stratos Four-Place Swinging Bucket Rotor

Catalog No. 76003026
Click to view available options
Capacity (Metric):
180 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Capacity (Metric) 180 mL
Capacity (English) 6 oz.
Includes Screw cap
Material Polypropylene
For Use With (Equipment) Stratos 4-place swinging bucket
Closure Type Screw Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.