Learn More
Thermo Scientific™ SUN-SRi™ Dissolution Flasks
Catalog No. 14823124
Change view
Click to view available options
Volume (Metric):
500 mL
900 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-823-124 500 mL
14-823-125 900 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-823-124 Supplier Thermo Scientific™ Supplier No. 100 011
Calibrated to contain and to deliver with permanently etched graduation lines and a write-on patch
  • Type 1, Class A borosilicate glass
  • Calibrated “to contain” and “to deliver” with permanently etched graduation lines and a write-on patch
  • Made specifically for the pharmaceutical industry
Compliance
Meets Federal Specifications NNN-F-289 and ASTM E288
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Volume (Metric) 500 mL