Learn More
Thermo Scientific™ SUN-SRi™ StepVial II™ Cap-Septa Kits
Description
Reduce chance of contamination while saving sample preparation time
- Caps are made of high quality polypropylene
- Closures have the profile of a crimp or snap closure for compatibility with robotic autosamplers
- Septa are 0.040 in. thick
Specifications
Specifications
| Material (Liner) | PTFE/Silicone |
| Color | Yellow |
| Material | Polypropylene |
| Pre-slit | No |
| Product Line | Surestop |