Learn More
Thermo Scientific™ Tuf-Bond™ PTFE/Silicone Discs
Catalog No. pi12720
Discs that combine the inertness of PTFE with the resealability of silicone
  • Structurally bonded PTFE to silicone; no cement to leak out of your sample after needle penetration
  • Reseals instantly, puncture after puncture
  • Autoclavable
  • Compresses to maintain a tight seal forcing the PTFE to conform to the sealing surface
  • Standard syringe and GC needles penetrate the entire disc with ease
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE, Silicone
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.