Learn More
Description
The Thermo Scientific TurboLuc Luciferase One-Step Glow Assay Kit measures TurboLuc16 (Tluc16) luciferase activity in mammalian cells with a single reagent-addition step, making it ideal for high-throughput screening (HTS) applications.
- Sensitive - extremely bright, Tluc16 luciferase allows detection at a single copy per cell
- Increased dynamic range - optimized reagents result in higher signal-to-noise ratio
- Stable signal - less than 40% signal loss over 60 min duration
- Convenient - one-step, mix-and-read, scalable protocol
- Automation-friendly - amenable to HTS protocols
- Simple - does not require a luminometer with injectors
Several features of the TurboLuc Luciferase One-Step Glow Assay Kit make it suitable for luminometers without injectors and for HTS applications. The convenient, one-step, homogenous protocol minimizes handling steps to support the use of automation. Simply combine the supplied substrate solution and assay buffer to make a single working solution, and then add it to microplate containing the transfected, treated cells. The stable signal output (glow enzyme kinetics) of this system provides flexibility with regard to lead time before read. When used with Thermo Scientific TurboLuc 16 luciferase vectors, the kit provides a highly sensitive bioluminescent reporter assay system for the detection of promoter or pathway activity.
Requires:
Tluc16 luciferase assay system and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan™ Ascent and Varioskan™ Flash microplate readers.
Applications:
- Gene reporter assays
- Promoter studies for analyzing cis- and trans-acting regulatory elements
- Drug screening
- Multiplexed assays to study off-target effects
- Cell signaling or signal transduction pathway analysis
Specifications
Specifications
| Type | Assay Kit |
| Target | Luciferase, TurboLuc™18 Luciferase |
| Sufficient For | 100 Reactions |
| Quantity | 100 Reactions |
| Content And Storage | TurboLuc One-Step Substrate (50X), 1 x 0.2 mL TurboLuc One-Step Assay Buffer, 11 mL Store in ultra-cold freezer (-68 to -85°C). |
| Assay | Reporter Enzyme, Luciferase Reporter Assay |
| Compatible Cells | Mammalian Cells |
| Detection Method | Bioluminescence |
| For Use With (Equipment) | Luminometer (Microplate) |
| Format | 384-well plate, 96-well plate |
| Show More |
Frequently Asked Questions (FAQs)
Unfortunately, we do not have an alternative product for the discontinued product TurboLuc Luciferase One-Step Glow Assay (Cat. No. 88263).
Here are possible causes and solutions:
- High luciferase expression: Reduce incubation time before collecting samples; Adjust instrument parameters to capture less signal (instrument-dependent); Decrease the amount of plasmid transfected into cells or decrease cell number.
- Control sample contaminated: Change pipette tips after each well.
Here are possible causes and solutions:
- Low transfection efficiency: Optimize transfection conditions using a visual transfection control (e.g., a plasmid over-expressing a fluorescent protein); Verify plasmid DNA quality using restriction digestion and agarose gel electrophoresis - use only transfection grade DNA; Note: Most high-quality plasmid DNA should be supercoiled; Use actively dividing, low passage cells; Use a different cell type.
- No promoter induction: Incubate cells under promoter-specific inducing conditions; Incubate the cells for a longer time after treatment; Change growth conditions to improve expression; Use a different promoter or cell type.
- TurboLuc One-Step Substrate auto-oxidized: Protect substrate from light and air and store at -80 degrees C; Prepare Working Solution immediately before use and protect from light.
- Wrong substrate used: Use only substrate supplied with the kit. Coelenterazine from related Luciferase kits (e.g., Renilla, Luciferase) will provide suboptimal performance.
- Low luciferase expression: Use a different promoter or growth conditions to improve expression; Adjust instrument parameters to capture more signal (instrument-dependent); Plate larger number of cells.
For Fetal Bovine Serum (FBS), there is no difference between 10% and 5% FBS. It is worthwhile to note that the type of serum does affect the luciferase assay. We tested the effect of 6 different types of sera (FBS, heat-inactivated FBS, dialyzed FBS, charcoal-treated FBS, donor adult bovine serum, and donor equine serum) on secreted luciferases, e.g. Gaussia, Gaussia-Dura, and Cypridina Luciferases. No noticeable difference was observed among different types of sera except for donor adult bovine serum. Unknown factors in donor adult bovine serum caused up to 35% inhibition in secreted luciferase activity. Therefore, we don’t recommend using donor adult bovine serum for Gaussia, Gaussia-Dura, and Cypridina Luciferases.
The problems with white plates are related to cross-talk blocking, surface reflectivity and tendency to phosphorescence. We do not recommend using white plates because they give much higher background even if adapted to darkness for 10 minutes. The white plastic does not block the light completely, and some portion of the signal will go through the plastic and will be seen when you measure adjacent wells. The typical situation is if you change from black to white plates, where the background is about doubled and the signal is increased about 10 times. Black plates are recommend for best signal to noise ratio even though the RLU values will be lower.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.