Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TurboLuc™ Luciferase One-Step Glow Assay Kit

Catalog No. p-4937579
Click to view available options
Quantity:
100 Reactions

Measure TurboLuc™16 (Tluc16) luciferase activity in mammalian cells with a single reagent-addition step that is for high-throughput screening (HTS) applications using the Thermo Scientific™ TurboLuc™ Luciferase One-Step Glow Assay Kit.

The Thermo Scientific TurboLuc Luciferase One-Step Glow Assay Kit measures TurboLuc16 (Tluc16) luciferase activity in mammalian cells with a single reagent-addition step, making it ideal for high-throughput screening (HTS) applications.

  • Sensitive - extremely bright, Tluc16 luciferase allows detection at a single copy per cell
  • Increased dynamic range - optimized reagents result in higher signal-to-noise ratio
  • Stable signal - less than 40% signal loss over 60 min duration
  • Convenient - one-step, mix-and-read, scalable protocol
  • Automation-friendly - amenable to HTS protocols
  • Simple - does not require a luminometer with injectors
This glow-type luciferase assay kit is designed for use with cultured mammalian cells expressing the Tluc16 luciferase reporter enzyme. Tluc16 is a 16 kDa, novel intracellular luciferase derived from the marine copedod Metridia luciferase family. The wild type luciferase has been modified to reduce its size, increase its brightness, enable its intracellular expression, and optimize it for HTS. A series of vectors with and without promoters are available for expression of Tluc16 luciferase reporter in mammalian cells.
Several features of the TurboLuc Luciferase One-Step Glow Assay Kit make it suitable for luminometers without injectors and for HTS applications. The convenient, one-step, homogenous protocol minimizes handling steps to support the use of automation. Simply combine the supplied substrate solution and assay buffer to make a single working solution, and then add it to microplate containing the transfected, treated cells. The stable signal output (glow enzyme kinetics) of this system provides flexibility with regard to lead time before read. When used with Thermo Scientific TurboLuc 16 luciferase vectors, the kit provides a highly sensitive bioluminescent reporter assay system for the detection of promoter or pathway activity.
Requires:
Tluc16 luciferase assay system and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan™ Ascent and Varioskan™ Flash microplate readers.
Applications:
  • Gene reporter assays
  • Promoter studies for analyzing cis- and trans-acting regulatory elements
  • Drug screening
  • Multiplexed assays to study off-target effects
  • Cell signaling or signal transduction pathway analysis

TRUSTED_SUSTAINABILITY

Specifications

Type Assay Kit
Target Luciferase, TurboLuc™18 Luciferase
Sufficient For 100 Reactions
Quantity 100 Reactions
Content And Storage TurboLuc One-Step Substrate (50X), 1 x 0.2 mL
TurboLuc One-Step Assay Buffer, 11 mL

Store in ultra-cold freezer (-68 to -85°C).
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
For Use With (Equipment) Luminometer (Microplate)
Format 384-well plate, 96-well plate
Label Type Enzyme Labeled
Substrate Properties Chemical Substrate
Technique Enhanced Chemiluminescence
Substrate Type Luciferase Substrate
Substrate Coelenterazine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.