Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ pMCS minP-Tluc16 Vector for Luciferase Assays
Click to view available options
:
pMCS minP-Tluc16 Vector for Luciferase Assays
Description
The Thermo Scientific pMCS Tluc16 Vectors are multiple cloning site (MCS) vectors designed to accept a promoter sequence for study of gene regulation using the intracellular TurboLuc™16 (Tluc16) luciferase reporter.
- Intracellular Tluc16 luciferase gene, optimized for high expression in mammalian systems
- Multiple cloning sites provide versatility for transfer of regulatory elements from one vector to another
- Synthetic polyA terminator and Transcriptional Pause Site (TPS) included upstream of MCS to minimize non-specific transcriptional read-through
- Dual-destabilization (DD) technology reduces accumulation of Tluc16 luciferase mRNA and protein in cells, enhancing the responsiveness of the assay
- Optimized minimal core promoter (minP) and 5' UTR region for efficient expression of Tluc16 luciferase
- Hygromycin resistance gene for use as selection marker to generate mammalian stable cell lines
- Prepared pNF-κB, and pCRE Tluc16-DD Vectors are designed to monitor the activation of NFκB protein and cAMP-binding protein (CREB) in mammalian cells
- Prepared pEF1α variety includes a modified version of the Elongated Factor 1α (EF1α) promoter for constitutive Tluc16 luciferase expression in mammalian cells

Specifications
Specifications
Type | Transcriptional Reporter Vector |
Cloning Method | Restriction Enzyme/MCS |
Delivery Type | Transfection |
Content And Storage | Store in freezer (-5 to -30°C). |
Reporter Gene | TurboLuc™16 Luciferase |
Product Line | Pierce™ |
For Use With (Application) | Reporter Assays |
Promoter | minP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction