Learn More
Thermo Scientific™ Ultrafast UFC-1 Phase GC Columns
Description
- Reduces analytical run time for increased throughput
- Long column lifetime
Specifications
Specifications
| Length (Metric) | 10 m |
| Film Thickness | 3 μm |