Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ UltraLink™ Iodoacetyl Micro Peptide Coupling Kit, 0.1 mL

Catalog No. pi20485
Change view
Click to view available options
Quantity:
1 Kit, 10 Columns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20485 1 Kit, 10 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20485 Supplier Thermo Scientific™ Supplier No. 20485

Covalently immobilize cysteine-peptides (sulfhydryl-containing molecules) for antibody affinity purification with these haloacetyl-activated polyacrylamide beads.

Thermo Scientific™ UltraLink Iodoacetyl Resin and Peptide Coupling Kit enable the covalent immobilization of cysteine-containing peptides for use in antibody purification and other affinity protocols.

The beaded UltraLink Iodoacetyl Resin is a hydrophilic copolymer of polyacrylamide and azlactone having a rigid polymeric structure with high surface area and pore volume. Each azlactone group has been modified to form a 15-atom spacer arm that terminates in an iodoacetyl group, which is capable of reacting with sulfhydryl groups (e.g., side chain of reduced cysteine residues) to covalently immobilize peptide or other sulfhydryl-containing ligands. The bead structure and efficiently coupling chemistry of Iodoacetyl-Activated UltraLink Biosupport results in high protein binding capacity, high linear flow rates, low nonspecific binding and overall superior performance in affinity chromatography. UltraLink Resins are ideal for medium pressure applications such as FPLC.

The Micro Peptide Coupling Kit is a microcentrifuge spin column kit for immobilizing small amounts (25 to 250μg) of sulfhydryl-containing peptides (e.g., cysteine-terminated peptides) onto the resin to create a small, reusable affinity column. The coupling and affinity purification procedures are optimized for small sample volumes (200 to 300μL). Wash and elution steps are achieved rapidly and efficiently with the convenient microcentrifuge spin columns. Each kit contains sufficient reagents for 10 coupling reactions and 20 affinity purifications. The kit is ideal for immobilizing peptide antigens that contain a terminal cysteine residue for use in purifying specific antibodies from small serum, ascites or culture supernatant samples.

Highlights:

  • High coupling efficiency and capacity – immobilizes sulfhydryl- containing proteins or other ligands with high efficiency and coupling capacity in one hour

  • Specific and leak-proof coupling chemistry – reacts specifically with sulfhydryl groups (reduced thiols), resulting in thioether bonds that are stable for use in many affinity purification procedures

  • Simple coupling conditions – perform immobilization reaction in any of a variety of buffers; coupling is most efficient and specific at pH 8.0 to 8.5; coupling compatible with or without organic solvent

  • Excellent reusability – prepared affinity resin can be used with typical binding and elution procedures for many cycles of affnity purification without significant loss of binding capacity

  • Durable, high-performance resin – porous beads have a 60μm diameter, can withstand 100 psi (6.9 bar) and allow for linear flow-through rates of 3000cm/hour

Micro Peptide Coupling Kit (Part No. 20485):

  • Optimized for small scale – ideal for coupling small amounts (25 to 250μg) of sulfhydryl-containing peptide or protein for purification of specific antibodies from crude serum

  • High-performance affinity resin – uses durable, polyacrylamide-based UltraLink Iodoacetyl Resin for specific reaction to sulfhydryl groups

  • Efficient coupling chemistry – immobilization efficiency >85% (as measured with 1 hour reaction using insulin, calcitonin and osteocalcin peptides)

  • Fast and easy to use – perform wash and elution steps using a microcentrifuge

  • Reusable – use prepared peptide resin several times with no significant loss of capacity

Includes:

Micro Peptide Coupling Kit contains microcentrifuge spin columns containing UltraLink Iodoacetyl Resin, coupling buffer, L-cysteine (for quenching), wash solution, PBS, elution buffer and collection tubes

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 25 to 250 μg of Peptide or Protein per Reaction
Description UltraLink Iodoacetyl Micro Peptide Coupling Kit
Content And Storage Sufficient For: Immobilizing 25–250 μg of peptide or protein per microcentrifuge column; Includes buffers for 20 affinity purifications
• UltraLink Iodoacetyl Spin Columns, 0.1 mL, 10 columns
• Coupling Buffer, 100 mL
• L-Cysteine·HCl, 100 mg
• Wash Solution, 25 mL
• PBS Pack (makes 500 mL), 1 pack
• IgG Elution Buffer, 50 mL
• Collection Tubes, 200 tubes
Store at 4°C protected from light.
Product Line UltraLink
Quantity 1 Kit, 10 Columns
Type Peptide Coupling Kit
Stationary Phase Haloacetyl
Sufficient For 10 Reactions
Format Kit, Microcentrifuge, Spin Column
Column Type Affinity, UltraLink™ Resin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.