Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ VersiCap Mat, 96-well, flat cap strips

Catalog No. p-4711145
Click to view available options
No. per Case:
25
100

Flexible mats of flat, domed cap strips for PCR or ultra clear cap strips for qPCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.

Thermo Scientific VersiCap Mats are unique, with each strip of eight caps being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.

The Thermo Scientific VersiCap Mats are unique, with each strip of eight caps being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.

Highlights

  • Multiple cap strips can be applied at the same time when sealing qPCR plates, thereby reducing setup time
  • Remove the need to cut up adhesive seals
  • Suitable for use with all Thermo Scientific 96-well PCR plates as well as standard and low profile strip tubes
Applications
  • PCR
  • qPCR/Real-Time PCR

Autoclavable Resistance: Successfully Tested
DMSO (100%) Resistance: Successfully Tested
Ethanol Resistance: Successfully Tested
Gamma Irradiation Resistance: Successfully Tested
Hazardous: No
Isopropanol Resistance: Successfully Tested
Long Term Storage: Successfully Tested
PCR (including waterbath): Successfully Tested
Peelable: Successfully Tested
Pierceable: Not Recommended
qPCR:Ultra Clear caps: Successfully Tested
Domed or Flat caps: Not Recommended
Resealable: Successfully Tested
Sealing Temperature Range: −20°C to +120°C
Shelf Life: 60 Months
Shipping Condition: Ambient
Storage Condition: Ambient
UV Irradiation Resistance: Successfully Tested

  • PCR
  • qPCR/Real-Time PCR

Certifications

Certified free from human DNA, RNases, and DNases

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Polypropylene
No. of Wells 96
No. per Case 25
Quantity 25 mats
Format Strips of 8 Flat Caps
Product Type PCR Plate Cap Strip
Product Line VersiPlate
Certified RNase/DNase-free Yes
Temperature Range (Metric) -20°C to +120°C
Plate Compatibility 96-well Plate
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.