Learn More
Thermo Scientific™ ViewPoint Enterprise Plug-Ins and Software
Description
Requires
System Specifications
Operates with Windows™ NT/2000/XP
Client Software Requirements
- Microsoft Windows™ NT 4 Service Pack 6 or
- Microsoft Windows™ 2000 Service Pack 4 or
- Microsoft Windows™ XP Service Pack 1 or higher
Plug-Ins
Each instrument, detector or sensor, which is capable of being remotely monitored by ViewPoint, has a unique Input Device Plug-In. This capability allows the instrument to communicate with ViewPoint Data Engine/System.
Client Plug-Ins
The ViewPoint User Interface and Data Engine are compatible with third-party Remote Monitoring Software systems. Compatability includes such systems as: MGPI-Teleview™, Industrial Innovative Solutions (IIS) – RADS™, and Bartlett – RMS™, and ESRI ArcView™ version 3.3 and 8. All purchased plug-ins are preloaded on the ViewPoint Appliance prior to delivery. Additional Input Device plug-ins, Client plug-ins, and User/Workstation License Bundles can be purchased and added to the original ViewPoint appliance via a patch file and updated security key
- Real-time personnel dosimetry and data management
- Area monitoring
- Integrated remote monitoring
- Instrument management
- First Responder environmental monitoring
- Site Process Monitoring
Specifications
Specifications
| Product Type | Viewpoint Enterprise Plug-Ins |
| Description | Third-party; Sold each |
| Operating System | Windows™ NT/2000/XP |
| For Use With (Application) | Real time personnel dosimetry and data management, area monitoring, integrated remote monitoring, instrument management, first responder environmental monitoring, site process monitoring |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.