Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vernier™ Thermocouple



A simple and rugged sensor designed to measure temperature in the range of –200ºC to 1400ºC. Low cost and a wide temperature range make the Thermocouple an economical, versatile temperature sensor.
Supplier: Vernier™ TCABTA
Includes: Vernier Thermocouple
Requires : Requires an interface and software, sold separately
Description
Typical Accuracy: 0ºC to 900ºC: ±2.2º –200ºC to 0ºC: ±5ºC 900ºC to 1400ºC: ±15ºC Power: 11 mA @ 5 VDC Typical Resolution: 0.40ºCWarranty and Services
5-year limited warranty
Specifications
Middle School, High School, College | |
5 Year Limited | |
0.40°C |
Required | |
Educational use only | |
11 mA at 5 VDC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction