Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thermus thermophilus cas3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117525
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117525 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117525 Supplier Invitrogen™ Supplier No. PA5117525
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

CRISPR (clustered regularly interspaced short palindromic repeat), is an adaptive immune system that provides protection against mobile genetic elements (viruses, transposable elements and conjugative plasmids). CRISPR clusters contain sequences complementary to antecedent mobile elements and target invading nucleic acids. CRISPR clusters are transcribed and processed into CRISPR RNA (crRNA). Cas3 plus Cascade participate in CRISPR interference, the third stage of CRISPR immunity. The N-terminal domain (residues 6-260) acts as a ssDNA endonuclease, has no activity on dsDNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thermus thermophilus cas3
Applications Western Blot
Classification Polyclonal
Concentration 4.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ygcB
Gene Accession No. P38036, Q53VY2
Gene Alias CRISPR-associated endonuclease; helicase Cas3
Gene Symbols cas3, ygcB
Host Species Rabbit
Immunogen Recombinant Thermus thermophilus CRISPR-associated endonuclease/helicase Cas3 protein (120-226aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3167883, 947229
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.