Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiamine Hydrochloride, Powder, MP Biomedicals

Catalog No. ICN10302825 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10302825 25 g
ICN10302880 100 g
ICN10302883 250 g
ICN10302891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10302825 Supplier MP Biomedicals, Inc Supplier No. 0210302825
Add to Cart
Add to Cart

Thiamine (Vitamin B1) is one of the essential vitamins and is required for carbohydrate metabolism.

Thiamine hydrochloride is used as a food-additive to add a brothy/meaty flavor to gravies or soups. It is also used as food supplement. Used in fluorometric determination of mercury.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H17ClN4OS·HCl
Molecular Weight (g/mol) 337.263
MDL Number MFCD00012780
Synonym Aneurine hydrochloride, Vitamin B1 hydrochloride, Vitamin B1 HCl, vitamin b1 mononitrate, aneurine mononitrate, aneurine nitrate, betabion mononitrate, thiamine nitrate salt, vitamin b sub 1 nitrate, thiamine mononitrate usp

Specifications

Quantity 25 g
Beilstein 3851771
IUPAC Name 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-
Formula Weight 337.3
Physical Form Powder
Chemical Name or Material Thiamine Hydrochloride, Food Supplement
EINECSNumber 200-641-8
storageNote1 Store Desiccated
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.