Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiamine Mononitrate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607369 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-369 1 kg Amber Glass Bottle
18-607-366 25 g Amber Glass Bottle
18-607-367 100 g Amber Glass Bottle
18-607-368 500 g Amber Glass Bottle
18-607-370 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-607-369 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TH1171KG
Add to Cart
Add to Cart

TH117, 532-43-4, C12H17N5O4S

Spectrum™ Chemical Thiamine Mononitrate, USP is used to treat or prevent vitamin B1 deficiency. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 532-43-4
Molecular Formula C12H17N5O4S
Molecular Weight (g/mol) 327.36
MDL Number MFCD00036330
InChI Key UIERGBJEBXXIGO-UHFFFAOYSA-N
IUPAC Name 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-3H-1λ4,3-thiazol-1-ylium nitrate
SMILES [O-][N+]([O-])=O.CC1=C(CCO)[S+]=CN1CC1=CN=C(C)N=C1N

Specifications

CAS Max % 1
pH 6.0 to 7.5
Ignition Residue 0.002
Loss on Drying 0.01
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Thiamine Mononitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.