Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thianaphthene, 97%

Catalog No. p-3737541
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 95-15-8 | C8H6S | 134.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-15-8
Molecular Formula C8H6S
Molecular Weight (g/mol) 134.2
MDL Number MFCD00005864
InChI Key FCEHBMOGCRZNNI-UHFFFAOYSA-N
Synonym benzo b thiophene, thianaphthene, benzothiophene, benzothiofuran, thionaphthene, 1-thiaindene, thianaphthen, benzothiophen, thianaphtene, 1-benzothiophen
PubChem CID 7221
ChEBI CHEBI:35858
IUPAC Name 1-benzothiophene
SMILES C1=CC=C2C(=C1)C=CS2

Specifications

Melting Point 28.0°C to 32.0°C
Color Pink to White or Yellow
Density 1.1400g/mL
Boiling Point 221.0°C to 222.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 17, 59
Specific Gravity 1.14
Merck Index 15, 9455
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in usual organic solvents, soluble in ethanol, ethylether, acetone
Formula Weight 134.2
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material Thianaphthene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
EINECSNumber 202-395-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.