Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiazole-5-carboxaldehyde, 95%, Thermo Scientific™

Catalog No. AAH6013803
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-32-3
Molecular Formula C4H3NOS
Molecular Weight (g/mol) 113.13
MDL Number MFCD02179516
InChI Key ZXRLWHGLEJGMNO-UHFFFAOYSA-N
Synonym thiazole-5-carboxaldehyde, thiazole-5-carbaldehyde, 5-thiazolecarboxaldehyde, thiazole-5-carboxyaldehyde, 5-formylthiazole, 5-formyl-1,3-thiazole, 1,3-thiazole-5-carboxaldehyde, thiazole-5-aldehyde, zlchem 512, pubchem14219
PubChem CID 2773577
IUPAC Name 1,3-thiazole-5-carbaldehyde
SMILES O=CC1=CN=CS1

Specifications

Density 1.304
Boiling Point 92°C to 98°C (16 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.5874
Quantity 1 g
Formula Weight 113.14
Percent Purity 95%
Chemical Name or Material Thiazole-5-carboxaldehyde
Hazard Category H302-H317-H319
Hazard Statement GHS H Statement
H301-H319-H317
Toxic if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.