Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THIK-2 Mouse anti-Mouse, Rat, Clone: N468/37, MilliporeSigma™

Catalog No. MABN242325 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN242325 25 μg
MABN2423100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN242325 Supplier MilliporeSigma Supplier No. MABN242325UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

THIK-2 Monoclonal antibody specifically detects THIK-2 in Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen THIK-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone N468/37
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen A recombinant fragment corresponding to 106 amino acids from the C-terminal, cytoplasmic domain of mouse THIK-2 (MNTK1).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.