Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thimet Oligopeptidase Monoclonal Antibody (455710)
GREENER_CHOICE

Catalog No. PIMA524174
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524174 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524174 Supplier Invitrogen™ Supplier No. MA524174
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human Neurolysin is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

A steady-state level of amyloid beta-peptide (Abeta) represents a balance between its biosynthesis from the amyloid precursor protein (APP) through the action of the beta and gamma-secretases and its catabolism by a variety of proteolytic enzymes. Thimet Oligopeptidase (THOP1) has been shown to promote A-beta degradation. It has also been shown to play a role in antigen presentation, metabolism of neuropeptides under 20 amino acids, and cytoplasmic peptide degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thimet Oligopeptidase
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 455710
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Thop1
Gene Accession No. P52888
Gene Alias AI131655; AI327041; EC 3.4.24; EC 3.4.24.15; endo-oligopeptidase A; Endopeptidase 24.15; EP24.15; MEPD_HUMAN; metalloendopeptidase; MP78; PZ-peptidase; soluble metallo-endopeptidase; Thimet oligopeptidase; thimet oligopeptidase 1; Thop1; TOP
Gene Symbols Thop1
Host Species Mouse
Immunogen E. coli-derived recombinant human Thimet Oligopeptidase/THOP1 Lys2-Cys689.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7064
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.