Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thimet Oligopeptidase Monoclonal Antibody (4D6)
GREENER_CHOICE

Catalog No. PIMA524783
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524783 Supplier Invitrogen™ Supplier No. MA524783
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thimet Oligopeptidase Monoclonal Antibody for Western Blot, IHC (P), ICC/IF

A steady-state level of amyloid beta-peptide (Abeta) represents a balance between its biosynthesis from the amyloid precursor protein (APP) through the action of the beta and gamma-secretases and its catabolism by a variety of proteolytic enzymes. Thimet Oligopeptidase (THOP1) has been shown to promote A-beta degradation. It has also been shown to play a role in antigen presentation, metabolism of neuropeptides under 20 amino acids, and cytoplasmic peptide degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thimet Oligopeptidase
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4D6
Concentration 1.54 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% sodium azide
Gene Thop1
Gene Accession No. P24155, P52888, Q8C1A5
Gene Alias AI131655; AI327041; EC 3.4.24; EC 3.4.24.15; endo-oligopeptidase A; Endopeptidase 24.15; EP24.15; MEPD_HUMAN; metalloendopeptidase; MP78; PZ-peptidase; soluble metallo-endopeptidase; Thimet oligopeptidase; thimet oligopeptidase 1; Thop1; TOP
Gene Symbols Thop1
Host Species Mouse
Immunogen A synthetic peptide corresponding to residues 56-72 of human Thimet Oligopeptidase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50492, 64517, 7064
Target Species Human, Mouse, Monkey, Rabbit, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.