Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thimet Oligopeptidase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597152
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597152 Supplier Thermo Scientific Supplier No. PA597152
Add to Cart
Add to Cart

Thimet Oligopeptidase Polyclonal Antibody

Thimet Oligopeptidase Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Thimet Oligopeptidase
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Thop1
Gene Accession No. P24155, P52888, Q8C1A5
Gene Alias AI131655; AI327041; EC 3.4.24; EC 3.4.24.15; endo-oligopeptidase A; Endopeptidase 24.15; EP24.15; MEPD_HUMAN; metalloendopeptidase; MP78; PZ-peptidase; soluble metallo-endopeptidase; Thimet oligopeptidase; thimet oligopeptidase 1; Thop1; TOP
Gene Symbols Thop1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human THOP1 (NP_0032401).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50492, 64517, 7064
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.