Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thimet Oligopeptidase/THOP1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298355APC
Description
Thimet Oligopeptidase/THOP1 Polyclonal antibody specifically detects Thimet Oligopeptidase/THOP1 in Mouse samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
Thimet Oligopeptidase/THOP1 | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
EC 3.4.24, EC 3.4.24.15, Endopeptidase 24.15, EP24.15, MEPD_HUMAN, MP78, thimet oligopeptidase, thimet oligopeptidase 1, TOP | |
Produced in rabbits immunized with purified, recombinant Mouse Thimet Oligopeptidase/THOP1 (Accession#: NP_073144.3; Lys 2-Cys687) | |
0.1 mL | |
Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
7064 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction