Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Thin Layer Chromatography Kit
GREENER_CHOICESCI_ED

Catalog No. s96897 Shop All Aldon Products
Click to view available options
Class Size:
15 Lab Groups

WARNING: This product can expose you to chemicals including Methanol/Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.

CHOKING HAZARD - Small parts. Not for children under 3 yrs.

WARNING: This product can expose you to chemicals including Methanol/Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.

CHOKING HAZARD - Small parts. Not for children under 3 yrs.

Includes

Materials for 15 groups: Food coloring set with 0.5mL each of yellow, red, blue and green; 30 mL Chromatography Solvent #1, water (5); 30 mL Chromatography Solvent #2, 50% Ethanol UN1170 (5); 30 mL Chromatography Solvent #3, 95% Ethanol UN1170 (5); thin-layer chromatography plates (15); and capillary pipets (4)

Requires

Plastic wrap, paraffin or tinfoil and beakers/containers large enough to hold the TLC plates

See that commercial food colors contain more than meet the eye.

  • Perform a highly effective separation procedure on four food colors
  • Determine if there is more in each color than visual appearance would suggest

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Perform a Highly Effective Separation Procedure on Four Food Colors
Includes Food coloring sample set containing 1 x 0.5 mL yellow, 1 x 0.5 mL red, 1 x 0.5 mL blue, 1 x 0.5 mL green, 5 x 30 mL chromatography solvent, 5 x 30 mL chromatography solvent, 5 x 30 mL chromatography solvent, 15 tlc sheets, 4 capillary pipettes
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type Thin Layer Chromatography Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.