Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioanisole, 99%

Catalog No. p-7022199
Click to view available options
Quantity:
25 g
100 g
500 g
2500 g

CAS: 100-68-5 | C7H8S | 124.201 g/mol

Thioanisole is used as an intermediate in the preparation of dyes, pharmaceuticals and agrochemicals. It is also used to prepare stable sulfonium salts, sulfoxides and sufones. It is utilized for the cleavage of methyl ethers in association with triflic acid. It acts as a flavoring agent or an adjuvant. It is an important starting material for the synthesis of 3-substituted benzo[b]thiophenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thioanisole is used as an intermediate in the preparation of dyes, pharmaceuticals and agrochemicals. It is also used to prepare stable sulfonium salts, sulfoxides and sufones. It is utilized for the cleavage of methyl ethers in association with triflic acid. It acts as a flavoring agent or an adjuvant. It is an important starting material for the synthesis of 3-substituted benzo[b]thiophenes.

Solubility
Miscible with most common organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-68-5
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.201
MDL Number MFCD00008559
InChI Key HNKJADCVZUBCPG-UHFFFAOYSA-N
Synonym thioanisole, methyl phenyl sulfide, methylthio benzene, thioanisol, benzene, methylthio, phenylthiomethane, phenyl methyl sulfide, methylsulfanyl benzene, methylphenylsulfide, 1-thiaethyl benzene
PubChem CID 7520
IUPAC Name methylsulfanylbenzene
SMILES CSC1=CC=CC=C1

Specifications

Melting Point -15°C
Density 1.058
Boiling Point 188°C to 189°C
Flash Point 73°C (163°F)
Odor Stench
Refractive Index 1.586
Quantity 25 g
UN Number UN1993
Beilstein 1904179
Solubility Information Miscible with most common organic solvents. Immiscible with water.
Formula Weight 124.21
Percent Purity 99%
Chemical Name or Material Thioanisole
Show More Show Less
Hazard Category H227-H302-H315-H317-H319
Hazard Statement GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 202-878-2
RTECSNumber DA6200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.