Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thiobenzamide 98.0+%, TCI America™

Click to view available options
Quantity:
25 g
100 g
Specifications
Specifications
Melting Point | 117°C |
Color | Yellow |
Quantity | 100 g |
UN Number | 2811 |
Formula Weight | 137.20 |
Percent Purity | ≥98.0% (HPLC,N) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Thiobenzamide |
Safety and Handling
RTECSNumber | CV5860000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction