Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiocarbanilide, 98%

Catalog No. AC138700010
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Plastic Drum
Plastic bottle

CAS: 102-08-9 | C13H12N2S | 228.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-08-9
Molecular Formula C13H12N2S
Molecular Weight (g/mol) 228.31
MDL Number MFCD00004921
InChI Key FCSHMCFRCYZTRQ-UHFFFAOYSA-N
Synonym thiocarbanilide, diphenylthiourea, 1,3-diphenyl-2-thiourea, n,n'-diphenylthiourea, sulfocarbanilide, sym-diphenylthiourea, thiourea, n,n'-diphenyl, stabilisator c, rhenocure ca, vulkacit ca
PubChem CID 700999
IUPAC Name 1,3-diphenylthiourea
SMILES S=C(NC1=CC=CC=C1)NC1=CC=CC=C1

Specifications

Melting Point 151.0°C to 154.0°C
Color White to Yellow
Loss on Drying 0.3% max. (60°C)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H5NHCSNHC6H5
Quantity 1 kg
Beilstein 12,394
Merck Index 15,3373
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Formula Weight 228.32
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Thiocarbanilide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 203-004-2
RTECSNumber FE1225000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.