Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiodiglycolic anhydride, 98%

Catalog No. AAA1339014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1339014 25 g
AAA1339006 5 g
AAA1339022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1339014 Supplier Thermo Scientific Chemicals Supplier No. A1339014
Add to Cart

CAS: 3261-87-8 | C4H4O3S | 132.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3261-87-8
Molecular Formula C4H4O3S
Molecular Weight (g/mol) 132.133
MDL Number MFCD00051689
InChI Key RIIUAPMWDSRBSH-UHFFFAOYSA-N
Synonym thiodiglycolic anhydride, 1-oxa-4-thia-cyclohexan-2,6-dione
PubChem CID 137871
IUPAC Name 1,4-oxathiane-2,6-dione
SMILES C1C(=O)OC(=O)CS1

Specifications

Melting Point 95°C to 100°C
Beilstein 112528
Formula Weight 132.13
Percent Purity 98%
Quantity 25 g
Sensitivity Moisture sensitive
Chemical Name or Material Thiodiglycolic anhydride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.