Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Thiol Peroxidase Protein
SDP

Catalog No. NBP17880510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17880510 100μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17880510 Supplier Novus Biologicals™ Supplier No. NBP178805100UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-168 of E.coli Thiol Peroxidase The Recombinant E. coli Thiol Peroxidase Protein is derived from E. coli. The Recombinant E. coli Thiol Peroxidase Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_415840
Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2mM DTT, 0.1M NaCl.
Molecular Weight (g/mol) 19.9kDa
Purification Method SDS-PAGE
Quantity 100μg
Source Recombinant E. coli TPX protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen TPX, 1-168aa, E. coli, His tag, E. coli (NP_415840).
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Thiol Peroxidase
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.