Learn More
Thionyl chloride, 1M soln. in dichloromethane, Thermo Scientific Chemicals
Description
Sulfuryl chloride and thionyl chloride have been found to be new chlorinating agents for alkyl sulfides. Preparation of aromatic polyesters by direct polycondensation with thionyl chloride in pyridine. Conversion of aromatic and. alpha.,. beta.-unsaturated aldehydes to dichlorides by thionyl chloride and dimethylformamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Molecular Formula | Cl2OS |
Linear Formula | SOCl2 |
UN Number | UN2922 |
Concentration | 1M soln. in dichloromethane |
Density | 1.35 |
Synonym | thionyl chloride, sulfinyl chloride, sulfurous dichloride, thionylchloride, sulfurous oxychloride, sulfur oxychloride, sulfinyl dichloride, socl2, dichlorosulfoxide, sulfur chloride oxide |
Solubility Information | Reacts with water. |
InChI Key | FYSNRJHAOHDILO-UHFFFAOYSA-N |
SMILES | O=S(Cl)Cl |
IUPAC Name | thionyl dichloride |
Show More |
Safety and Handling
Hazard Category | H314-H335-H350 |
Hazard Statement | GHS H Statement H314-H351 Causes severe skin burns and eye damage. Suspected of causing cancer. |
Precautionary Statement | P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S. |
EINECSNumber | 231-748-8 |
RTECSNumber | XM5150000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.