Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Thioperamide
GREENER_CHOICESDP

Catalog No. 064450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-445-0 50 mg
06-441-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 06-445-0 Supplier R&D Systems™ Supplier No. 0644/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. H3 antagonist; also H4 inverse agonist

Potent histamine H3 and H4 antagonist/inverse agonist. Ki values are 25 and 27 nM for human recombinant H3 and H4 receptors respectively. Blocks eosinophil shape change (IC50 = 1.4 μM) and chemotaxis (IC50 = 519 nM) induced by histamine. Freely crosses the blood-brain barrier.

Chemical Identifiers

CAS 106243-16-7
Molecular Formula C19H28N4O4S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 292.44
Purity >99%
Molecular Weight (g/mol) 408.517
Chemical Name or Material Thioperamide
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.