Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophene-2,5-dicarboxylic acid, 97%

Catalog No. AAL1029303
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 4282-31-9 | C6H4O4S | 172.154 g/mol

Thiophene-2,5-dicarboxylic acid may be used in the synthesis new cobalt(II) thiophenedicarboxylate coordination polymer and novel mesogenic thiophene based supramolecular liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2,5-dicarboxylic acid may be used in the synthesis new cobalt(II) thiophenedicarboxylate coordination polymer and novel mesogenic thiophene based supramolecular liquid crystals.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4282-31-9
Molecular Formula C6H4O4S
Molecular Weight (g/mol) 172.154
MDL Number MFCD00016896
InChI Key YCGAZNXXGKTASZ-UHFFFAOYSA-N
Synonym 2,5-thiophenedicarboxylic acid, 2,5-dicarboxythiophene, unii-5q8a2g9y8a, 2,5-thiophene dicarboxylic acid, pubchem5580, maybridge4_002021, acmc-1aj5p, ksc182i5d, 2,5-thiophene-dicarboxylic acid, thiophene-2.5-dicarboxylic acid
PubChem CID 20273
IUPAC Name thiophene-2,5-dicarboxylic acid
SMILES C1=C(SC(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 355°C to 360°C
Quantity 1 g
Beilstein 136781
Solubility Information Very soluble in water.
Formula Weight 172.16
Percent Purity 97%
Chemical Name or Material Thiophene-2,5-dicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-284-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.