Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophene-2-acetonitrile 98.0+%, TCI America™
SDP

Catalog No. T079925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T079925ML 25 mL
T07995ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T079925ML Supplier TCI America Supplier No. T079925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20893-30-5
Molecular Formula C6H5NS
Molecular Weight (g/mol) 123.173
MDL Number MFCD00005453
InChI Key CLSHQIDDCJTHAJ-UHFFFAOYSA-N
Synonym 2-thiopheneacetonitrile, 2-thiophen-2-yl acetonitrile, 2-thienylacetonitrile, thiophene-2-acetonitrile, 2-cyanomethyl thiophene, 2-cyanomethylthiophene, 2-thiophene-acetonitrile, thiophen-2-yl-acetonitrile, thien-2-ylacetonitrile, thiophen-2-ylacetonitrile
PubChem CID 72880
ChEBI CHEBI:27382
IUPAC Name 2-thiophen-2-ylacetonitrile
SMILES C1=CSC(=C1)CC#N

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 mL
UN Number 3276
Formula Weight 123.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Thiophene-2-acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.