Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophene-2-thiol, contains dimer, 97% as monomer

Catalog No. AAB2264203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2264203 1 g
AAB2264206 5 g
AAB2264214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2264203 Supplier Thermo Scientific Chemicals Supplier No. B2264203
Only null left
Add to Cart

CAS: 7774-74-5 | C4H4S2 | 116.196 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7774-74-5
Molecular Formula C4H4S2
Molecular Weight (g/mol) 116.196
MDL Number MFCD00051666
InChI Key SWEDAZLCYJDAGW-UHFFFAOYSA-N
Synonym 2-mercaptothiophene, 2-thiophenethiol, thiophenethiol, 2-thienyl mercaptan, 2-thienylthiol, 2-thiophene thiol, unii-f3lb1437xq, 2-sulphanylthiophene, alpha-thienyl mercaptan, thiophene thiol
PubChem CID 522674
IUPAC Name thiophene-2-thiol
SMILES C1=CSC(=C1)S

Specifications

Density 1.247
Boiling Point 62°C to 65°C (12 mmHg)
Flash Point 66°C (151°F)
Odor Unpleasant
Refractive Index 1.617
Quantity 1 g
Beilstein 104650
Sensitivity Air and light sensitive
Formula Weight 116.2
Percent Purity 97%
Assay as monomer, contains dimer
Chemical Name or Material Thiophene-2-thiol
Show More Show Less
Hazard Category H227-H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H227-H332-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-881-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.